Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GYPB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117102
Description
Positive Samples: Human serum Immunogen sequence: MYGKIIFVLL LSEIVSISAL STTEVAMHTS TSSSVTKSYI SSQTNGETGQ LVHRFTVPAP VVIILIILCV MAGIIGTILL ISYTIRRLIK A.
This protein is a minor sialoglycoprotein in erythrocyte membranes.
Specifications
GYPB | |
Polyclonal | |
Unconjugated | |
GYPB | |
CD235b; glycophorin B (MNS blood group); glycophorin B/glycophorin A fusion protein; glycophorin Hop; glycophorin-B; GPB; GYP; GYPB; GYPB/GYPA fusion; hybrid glycophorin Kip; MNS; PAS-3; Sialoglycoprotein delta; SS; Ss blood group; SS-active sialoglycoprotein | |
Rabbit | |
Affinity chromatography | |
RUO | |
2994 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.64 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P06028 | |
GYPB | |
A synthetic peptide corresponding to a sequence within amino acids 1-91 of human GYPB (NP_0020913). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction