Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GYPC Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 181471AP

Catalog No. 50-173-2013


Add to cart

Description

Description

This antibody is raised against the full-length protein of human Glycophorin-C. The apparent molecular weight of Glycophorin-C is lager than the calculated molecular weight due to glycosylation.

Glycophorin C (GYPC) is an integral membrane glycoprotein. It is a minor species carried by human erythrocytes, but plays an important role in regulating the mechanical stability of red cells. A number of glycophorin C mutations have been described. The Gerbich and Yus phenotypes are due to deletion of exon 3 and 2, respectively. The Webb and Duch antigens, also known as glycophorin D, result from single point mutations of the glycophorin C gene. The glycophorin C protein has very little homology with glycophorins A and B.
Specifications

Specifications

GYPC
0.41 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P04921
CD236, CD236R, GE, GLPC, Glycoconnectin, Glycophorin C, Glycophorin D, Glycoprotein beta, GPC, GPD, GYPC, GYPD, PAS 2, Sialoglycoprotein D
Rabbit
IgG
150 μL
-20°C
Primary
2995
Western Blot
Unconjugated
GYPC
Liquid
GYPC
GYPC Fusion Protein Ag12825
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit