Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Granzyme M Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549588
Description
Cleaves peptide substrates after methionine, leucine, and norleucine. Physiological substrates include EZR, alpha- tubulins and the apoptosis inhibitor BIRC5/Survivin. Promotes caspase activation and subsequent apoptosis of target cells.Specifications
Granzyme M | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P51124 | |
GZMM | |
KLH conjugated synthetic peptide between 84-113 amino acids from the Central region of human GZMM. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
GZMM | |
Granzyme M, 3421-, Met-1 serine protease, Hu-Met-1, Met-ase, Natural killer cell granular protease, GZMM, MET1 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
3004 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction