Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-2Dd Mouse anti-Mouse, BUV395, Clone: 34-2-12, BD Optibuild™

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 742466

Catalog No. BDB742466


Add to cart

Description

Description

The 34-2-12 antibody (also known as 34-2-12S) recognizes theα3 domain of the H-2D[d]. The binding of the antibody to its epitope is independent of theα1 andα2 domains andβ2 microglobulin. It cross-reacts with cells of the C3H.LG/Ckc strain. Reactivity with other haplotypes (eg, b, f, k, p, q, r, s /) has not been observed. Soluble mAb 34-2-12 blocks binding of the Ly-49A-expressing T lymphoma EL4 to immobilized H-2D[d]. However, further studies utilizing this mAb indicate that theα3 domain is not involved in the interaction between Ly-49A, or Ly-49G2, and H-2D[d].

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
Specifications

Specifications

H-2Dd
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Histocompatibility-2Dd, H-2Dd
(C57BL/6 x DBA/2)F1 mouse splenocytes
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV395 under optimal conditions that minimize unconjugated dye and antibody.
RUO
Primary
Murine
34-2-12
Brilliant Ultraviolet 395
Affinity Purified
Murine
Mouse C3H, also known as C3H/He, C3H/Bi IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit