Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H-2Dd Monoclonal Antibody (34-5-8S), PE, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen A15445
Description
H-2Dd Monoclonal antibody specifically detects H-2Dd in Mouse samples. It is validated for Flow Cytometry.Specifications
H-2Dd | |
Monoclonal | |
0.1 mg/mL | |
PBS with <0.1% sodium azide | |
P01895, P01896, P01897, P01899, P01900, P14426, P14427 | |
H2-D1 | |
mouse H-2Dd from BDF1 mouse splenocytes | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
34-5-8S | |
PE | |
H2-D1 | |
H-2 class I histocompatibility antigen, D-B alpha chain; H-2D; H2-D; H-2D cell surface glycoprotein; H-2D(B); H2D1; H2-D1; H2-DC1-beta transplantation antigen protein; H2DD; H2L; H2LD; histocompatibility 2, D region locus 1; MHC class I antigen; MHC class I H2 antigen; MHC class I H-2Dp; MHC H2-D-q alpha-chain; Q5k protein | |
Mouse | |
purified | |
RUO | |
14964 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction