Learn More
H-2Kk Mouse anti-Mouse, Alexa Fluor 700, Clone: 16-3-22S, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128202AF700
Catalog No. NBP128202AI
Add to cart
Description
Description
H-2Kk Monoclonal antibody specifically detects Antigen in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
H-2Kk
16-3-22S
Flow Cytometry
Purified
H2-K1
C3H mouse splenocytes
Protein A or G purified
RUO
Primary
14972
Mouse
Flow Cytometry
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
adrenomedullin binding protein, age-related maculopathy susceptibility 1, AHUS1, AMBP1, ARMD4, beta-1H, beta-1-H-globulin, CFHL3, complement factor H, factor H, factor H-like 1, FH, FHL1, H factor 1, H factor 1 (complement), H factor 2 (complement), HF, HF1ARMS1, HF2, HUSMGC88246
Mouse
IgG2a Kappa
0.25 ml
Store at 4C in the dark.
Monoclonal
An epitope on mouse H-2Kk MHC class I alloantigen