Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H-2Kk Monoclonal Antibody (36-7-5), FITC, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen MM4201
Description
H-2Kk Monoclonal antibody specifically detects H-2Kk in Mouse samples. It is validated for Flow Cytometry.Specifications
H-2Kk | |
Monoclonal | |
0.5 mg/mL | |
PBS with <0.1% sodium azide | |
P01901, P03991, P04223, P14428 | |
H2-K1 | |
Mouse H-2Dk. from A.AL mouse splenocytes | |
500 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
36-7-5 | |
FITC | |
H2-K1 | |
class I major histocompatibility antigen H-2Kd; H-2 class I histocompatibility antigen, K-B alpha chain; H-2 class I histocompatibility antigen, K-D alpha chain; H-2 class I histocompatibility antigen, K-K alpha chain; H-2 class I histocompatibility antigen, K-Q alpha chain; H-2 class I histocompatibility antigen, K-W28 alpha chain; H2DD; H2K; H-2K; H2-K; H-2K(B); H-2K(d); H-2K(K); H-2K(Q); H2K1; H2-K1; H2KD; H2KK; histocompatibility 2, K1, K region; K-f; MHC class I antigen; MHC class I H2-K-b-alpha-2 cell surface glycoprotein; MHC class I heavy chain H2-K; MHC H2-K transplantation antigen | |
Mouse | |
purified | |
RUO | |
14972 | |
4° C, store in dark | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction