Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-2Kk Mouse anti-Mouse, Phycoerythrin, Clone: 16-3-22S, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP128202PE

Catalog No. NBP128202PE



Description

Description

H-2Kk Monoclonal antibody specifically detects Antigen in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

H-2Kk
16-3-22S
Flow Cytometry
Purified
H2-K1
C3H mouse splenocytes
Protein A or G purified
RUO
Primary
14972
Mouse
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
adrenomedullin binding protein, age-related maculopathy susceptibility 1, AHUS1, AMBP1, ARMD4, beta-1H, beta-1-H-globulin, CFHL3, complement factor H, factor H, factor H-like 1, FH, FHL1, H factor 1, H factor 1 (complement), H factor 2 (complement), HF, HF1ARMS1, HF2, HUSMGC88246
Mouse
IgG2a Kappa
0.25 ml
Store at 4C in the dark.
Monoclonal
An epitope on mouse H-2Kk MHC class I alloantigen
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit