Learn More
Medchemexpress LLC H-9 dihydrochloride | 116700-36-8 | MFCD00069286 | 99.0% | 324.23 g/mol | C11H15Cl2N3O2S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1009235MG
Encompass_Preferred
H-9 dihydrochloride is the dihydrochloride salt of H-9, a small-molecule protein kinase A (PKA) inhibitor used in biochemical and cellular research to block PKA-mediated phosphorylation. Supplied as a dry solid, it has a molecular weight of 324.23 g/mol and is commonly prepared as DMSO stock solutions for in vitro and in vivo applications.
  • Inhibits protein kinase A activity in biochemical and cellular assays.
  • Applicable to studies of serotonin (5-HT) receptor and EGFR signaling pathways.
  • High purity suitable for research-grade experiments (≈99%).
  • Soluble in DMSO and compatible with common in vivo vehicles.
  • Store sealed and away from moisture to preserve stability.
Catalog No. 50-002-12387