Learn More
H-B Instrument™ SP Scienceware™ H-B™ FRIO-Temp™ Oven Verification Thermometers with Individual Calibration Report; Traceable to NIST
Verify sample temperatures in ovens with these mercury free, individually calibrated thermometers. H-B™ FRIO-Temp™ Oven Verification Thermometers Provide accurate temperature readings without sample contamination.
Manufacturer:  H-B Instrument™ B602001500
 View more versions of this product
Catalog No. 13-202-871
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Filled bottle (included) mimics the sample
  • PFA coated thermometers contain glass and liquid in case of breakage; filled with organic liquid
  • Permanently fused markings and lead free glass
  • Includes individual Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration
  • Individually serialized and made in the USA
Specifications
Specifications
2°C
200°C
Glass Beads
Yes
145 mm
±2°C; ±4°C above 130°C
50°C to 200°C
50°C
30 mL
100°C
Enviro-Safe™ Liquid
5.7 in.