Learn More
H-B Instrument™ Durac™ Alcohol Proof - Ethyl Alcohol Hydrometers
Used to determine proof of ethyl alcohol spirits, these hydrometers are available in various ranges from 0 to 206%
Manufacturer:  H-B Instrument Company B618074200
 View more versions of this product
Catalog No. 13-203-195
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
±0.2%
Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC /MRA and located in ISO 14001:2004 Environmental Management System registered facility.
Determining proof of Ethyl Alcohol Spirit
0.2%
300mm
0 to 20%
Safety and Handling
Recommended Storage : Alcohol proof hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.