Learn More
H-B Instrument™ Durac™ Brix Sugar Scale Plain Form Hydrometers
Measuring percent sucrose by weight, these plain form hydrometers are available in various ranges from -5° to +71° Brix
Manufacturer:  H-B Instrument Company B618020200
 View more versions of this product
Catalog No. 13-203-204
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
±0.1°
Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC/MRA and located in ISO 14001:2004 Environmental Management System registered facility.
Used in the food and beverage and other industries.
0.1°
300mm
10 to 20°
Safety and Handling
Recommended Storage : Plain form hydrometers should be stored in a clean, dry, safe place and should not be subjected to extreme temperatures.