Learn More
H-B Instrument™ Durac™ Galileo Thermometers
Thermometers with colorful spheres which rise and fall to indicate temperature
Manufacturer:  H-B Instrument Company B620001100
 View more versions of this product
Catalog No. 13-201-918
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
±2°C
7 in.
Glass
Not Applicable
Colorful spheres
177mm
2
18° to 26°C