Learn More
H-B Instrument™ Durac™ Isopropyl Alcohol Hydrometers
Measures the percent by volume of isopropyl alcohol
Manufacturer:  H-B Instrument Company B618094000
 View more versions of this product
Catalog No. 13-203-199
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
±1% by vol
Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC /MRA and located in ISO 14001:2004 Environmental Management System registered facility.
For measuring the percent by volume of isopropyl alcohol
1%
260mm
0 to 50%
Safety and Handling
Recommended Storage : Isopropyl alcohol hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.