Learn More
H-B Instrument™ SP Scienceware™ Durac™ Plus™ ASTM-Like Total Immersion Laboratory Thermometers
Designed for use in quality control laboratories or when mercury filled thermometers cannot be used, but ASTM standards are still needed. H-B Instrument - SP Scienceware™ Durac™ Plus™ ASTM-Like Total Immersion Laboratory Thermometers are safer to use than their mercury filled counterparts. 
Manufacturer:  H-B Instrument™ B607507000
 View more versions of this product
Catalog No. 13-202-874
Add to cart
Description
Description
  • Individually serialized
  • Blue organic fill thermometers
  • Lead-free white back glass
  • For use in quality control laboratories or where ISO™ 9000 standards are in place
  • Individually packaged
Specifications
Specifications
134°F
19F
120°F to 134°F
Quality control laboratories or where ISO 9000 standards are in place
280 mm
Blue Organic Liquid
±0.6°F
120°F
0.2°F
ASTM, ISO 9000
Total
Glass
ASTM-Like Total Immersion Laboratory Thermometers
11 in.