Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-B Instrument™ Durac™ Plus™ ASTM-Like Total Immersion Laboratory Thermometers

Safer than their mercury counterparts

Manufacturer:  H-B Instrument Company B607501400

 View more versions of this product

Catalog No. 13-201-713


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

0.2°C
ASTM, ISO 9000
Total
275mm
0.1°C
Blue Organic
17C
Quality control laboratories or where ISO 9000 standards are in place
10.8 in.
White Back Glass
19° to 27°C
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit