Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Durac™ Plus™ Certified Total Immersion Thermometers

With accuracy of 1.5°C or better.

Manufacturer:  H-B Instrument Company B602051400

 View more versions of this product

Catalog No. 13-201-627


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

1°C
Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries
18.1 in.
White Back Glass
-1° to +51°C
ISO/IEC 17025:2005, A2LA, NIST
Total
460mm
0.1°C
Organic
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit