Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-B Instrument™ Durac™ Plus™ Liquid-In-Glass PFA-Coated Total Immersion Thermometers

Provides safety and protection from breakage

Supplier:  H-B Instrument™ B607101400

 View more versions of this product

Catalog No. 13-201-615

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Includes: Statement of Traceability indicating accuracy

Description

Description

  • PFA safety coating keeps thermometer liquid and glass encapsulated in the event of breakage
  • Contain blue organic liquid against lead-free white back glass
  • Traceable to NIST, individually serialized
  • Include multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
  • Tested and inspected by our USA quality team
  • Coated thermometers are just as accurate as uncoated thermometers, but are not recommended for timed tests as the coating can slow reaction time

Certifications

NIST traceable

Specifications

Specifications

ISO/IEC 17025:2005, A2LA, NIST
Total Immersion
305 mm
Organic
0°C
±3°F; ±4°F above 212°F
Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries
12 in.
Glass
110°C
0°F to 230°F
2°F
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit