Learn More
H-B DURAC™ SP Scienceware™ Probeless Electronic Window Thermometers
Designed for use in high moisture or wet environments
Manufacturer:  H-B DURAC™ B609002500RMD
Catalog No. 13-201-953
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Requires : LR44 battery
Description
Description
  • Window thermometer (13-201-953) features easy installation with no wiring and is useful for cleanroom applications, fume hoods, glove boxes, greenhouses and any glass environment
  • Certifications
    CE marked
Specifications
Specifications
−10°C
±1°C
-10°C to +50°C
Window Temperature Measurement
44 mm
Water Resistant IP65; Requires LR44 Battery
50°C
0.1°C
1.7 x 5 x 1.7 in. (44 x 127 x 44 mm)
1.7 in.
0.1°C/°F