Learn More
H-B Instrument™ Durac™ Plus™ Ultra Low Thermometers
Measures ultra low temperatures
Manufacturer:  H-B Instrument™ B607600600
 View more versions of this product
Catalog No. 13-203-021
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Filled with Toluene
  • White back glass
  • Meniscus visible in appropriate low temperature application
  • Individually packaged
Low temperatures may cause dyes to separate.
Compliance
Meets all requirements for physical dimensions and accuracy of ANSI/SAMA Z236.1-1983 (Total immersion).
Specifications
Specifications
-50°C to +50°C
50°C
Total
300 mm
2°C
−50°C
12 in.
Glass