Learn More
H-B Instrument™ Durac™ Plus™ Ultra Low Thermometers
Measures ultra low temperatures
Specifications
Length (English) Overall 12 in.
Material Glass
View More Specs
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
13-203-021
View Documents
H-B Instrument™
B607600600
N/A Total Immersion; Range: -50° to +50°C; Divisions: 1; Accuracy: 2°C This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Filled with Toluene
  • White back glass
  • Meniscus visible in appropriate low temperature application
  • Individually packaged
Low temperatures may cause dyes to separate.
Compliance
Meets all requirements for physical dimensions and accuracy of ANSI/SAMA Z236.1-1983 (Total immersion).
Specifications
Specifications
12 in.
Glass