H-B™ Durac™ Specific Gravity/Relative Density Plastic Plain Form Hydrometers

Shatterproof polycarbonate prevents contamination

Manufacturer: H-B Instrument Company B618000100

 View more versions of this product

Catalog No. 13-201-851

  Check Availability
Add to Cart

Alert

Polycarbonate hydrometers are not recommended for use with petroleum-based liquids or other corrosives that would potentially affect the plastic material. They should be stored in a clean, dry, safe place and should not be subjected to extreme temperatures.


    Specifications

    For Use With (Application) Ideal in situations where rough handling or contamination could be an issue, such as in the food or beverage industries.
    Specific Gravity Range 0.920 to 1.080
    Accuracy ±0.002
    Increments 0.002
    Certifications/Compliance Hydrometers are made and hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC /MRA and located in ISO 14001:2004 Environmental Management System registered facility. They are packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST.
    Length (Metric) 300mm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit