Learn More
H-B Instrument™ Durac™ Rain Guage
Nesting ability enables easy use and storage
Manufacturer:  H-B Instrument™ B615070000RMD
Catalog No. 13-202-009
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Rainfall chart
Description
Description
  • Cylindrical construction of three pieces with inner and outer removable cylinders for collecting rain and rain overflow
  • Inner cylinder features funnel of 100mm diameter with dual scale that shows both millimeter and inch markings
  • Plastic construction withstands elements and makes it durable
Specifications
Specifications
11.5 in.
29.2cm
Rainfall chart
Durac rain gauge