Learn More
H-B Instrument™ Durac™ Special Application Thermometers
Individually serialized red spirit thermometer liquid
Specifications
Immersion Type Partial
Thermometer Fill Red Organic Liquid
Material Glass
View More Specs
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
13-201-557
View Documents
H-B Instrument™
B608102600
N/A Divisions: 1°; Accuracy: 1°C; Temp. range: 24° to 57°C; Length: 225mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Special Application Thermometers, Incubator
  • Designed with swell in the body holding thermometer in place
  • Glass swell
  • Durable and accurate
  • Individually packaged in recyclable Safety-Pak™ packaging with a non-roll fitting
Special Application Thermometers, Dry Block
  • Designed for use in all standard heating blocks and water baths
  • Individually packaged in recyclable packaging
Specifications
Specifications
Partial
Red Organic Liquid
Glass