H-B Instrument™ Durac™ ASTM Precision Specific Gravity Hydrometers with Individual Calibration Report: for Heavy Liquids

Available in a variety of individually calibrated models measuring specific gravity of heavy liquids in various ranges from 0.650 to 1.500

Manufacturer: H-B Instrument Company B618981800

 View more versions of this product

Catalog No. 13-203-056

  Check Availability
Add to Cart

    Specifications

    Recommended Storage ASTM calibrated hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.
    Calibration Points 1.005, 1.025, 1.045
    For Use With (Application) For Heavy Liquids.
    Accuracy ±0.0005
    Certifications/Compliance Calibrated and hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC/MRA and located in ISO 14001:2004 Environmental Management System registered facility. Conforms with American Society of Testing Materials (ASTM) E100 standards.
    Specific Gravity Range 1.000 to 1.050
    ASTM Number 111H
    Increments 0.0005
    Length (Metric) 330mm

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit