Learn More
H-B Instrument™ Durac™ ASTM Precision Specific Gravity Hydrometers with Individual Calibration Report: for Heavy Liquids
Available in a variety of individually calibrated models measuring specific gravity of heavy liquids in various ranges from 0.650 to 1.500
Manufacturer:  H-B Instrument Company B618982400
 View more versions of this product
Catalog No. 13-203-060
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
115H
0.0005
330mm
1.200 to 1.250
±0.0005
Calibrated and hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC/MRA and located in ISO 14001:2004 Environmental Management System registered facility. Conforms with American Society of Testing Materials (ASTM) E100 standards.
For Heavy Liquids.
1.200, 1.225, 1.245
Safety and Handling
Recommended Storage : ASTM calibrated hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.