Learn More
H-B Instrument™ Durac™ Specific Gravity ASTM Hydrometers: for Heavy Liquids
Available in a variety of models measuring specific gravity of heavy and light liquids in various ranges from 0.650 to 2.000
Manufacturer:  H-B Instrument Company B618911800
 View more versions of this product
Catalog No. 13-203-069
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
114H
0.0005
330mm
1.150 to 1.200
±0.0005
Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC/MRA and located in ISO 14001:2004 Environmental Management System registered facility. Conforms to American Society of Testing Materials (ASTM) E100 standards.
Commonly used in industries such as food and beverage, chemical, pharmacology, transportation, refrigeration and more.
For Heavy Liquids
Safety and Handling
Recommended Storage : ASTM hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.