Learn More
H-B Instrument™ Durac™ Specific Gravity ASTM Hydrometers: for Light Liquids
Available in a variety of models measuring specific gravity of heavy and light liquids in various ranges from 0.650 to 2.000
Manufacturer:  H-B Instrument Company B618911400
 View more versions of this product
Catalog No. 13-203-108
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
108H
0.001
260mm
0.950 to 1.000
±0.001
Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC/MRA and located in ISO 14001:2004 Environmental Management System registered facility. Conforms to American Society of Testing Materials (ASTM) E100 standards.
Commonly used in industries such as food and beverage, chemical, pharmacology, transportation, refrigeration and more.
Safety and Handling
Recommended Storage : ASTM hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.