Learn More
H-B Instrument™ Durac™ Specific Gravity/Relative Density (g/cm3) Plain Form Hydrometers: for Liquids Heavier Than Water
Available in a variety of models measuring specific gravity/relative density (g/cm3) of liquids heavier than water in various ranges from 0.690 to 3.000
Manufacturer:  H-B Instrument Company B618017100
 View more versions of this product
Catalog No. 13-202-119
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
±0.005
Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory (Cert 2448.01); internationally recognized and accepted by the signatory economies of the ILAC/MRA and located in ISO 14001:2004 Environmental Management System registered facility.
For Liquids Heavier Than Water
0.005
300mm
1.500 to 2.000
Safety and Handling
Recommended Storage : Plain form hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.