Learn More
H-B Instrument™ Durac™ Specific Gravity/Relative Density (g/cm3) Precision Hydrometers
Available in a wide range of models measuring specific gravity/relative density (g/cm3) of a variety of liquids in various ranges from 0.600 to 0.920
Manufacturer:  H-B Instrument Company B618011900
 View more versions of this product
Catalog No. 13-202-085
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
±0.001
Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC/MRA and located in ISO 14001:2004 Environmental Management System registered facility.
Specific gravity hydrometers are the most widely used hydrometer, common in industries such as food and beverage, chemical, pharmacology, transportation, refrigeration and more.
0.0005
325mm
1.720 to 1.790
Safety and Handling
Recommended Storage : Precision hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.