Learn More
H-B Instrument™ Durac™ Thermometer-Hygrometers
For measurement of temperature and humidity
Manufacturer:  H-B Instrument™ B615050100
 View more versions of this product
Catalog No. 13-201-845
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Comes with loop hole for wall mounting (Cat Nos. 235C and 235F)
  • Ideal for use with environments that can withstand the elements such as inside incubators, refrigerators and freezers (Cat Nos. 3725C and 3725F)
  • Available in two materials stainless-steel and plastic
  • With accuracy of 2°C; 4% RH
Specifications
Specifications
2°F, 4% RH
12.7 cm
1°F, 2% RH
32°F to 248°F
0°C to 120°C
5 in.
Measurement of temperature and humidity, ideal for use with environments that can withstand the elements such as inside incubators, refrigerators and freezers
Stainless Steel
0 to 100%
Thermohygrometer