H-B Instrument™ Durac™ Waterproof Electronic Thermometer

For reading temperatures in sample reagents, water baths, semi-solids and gases

Manufacturer: H-B Instrument Company B609001000RMD

 View more versions of this product

Catalog No. 13-201-769

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ISO/IEC 17025:2005, A2LA, NIST
    Accuracy 2°C
    For Use With (Equipment) Field studies, cuvettes, test tubes, flasks and beakers, reads temperature in sample reagents, water baths, semi solids and gases
    Includes Stainless Steel probe, probe cover with pocket clip, multilanguage instructions and statement of accuracy, CR2032 battery
    Length (English) 3 in.
    Length (English) Probe 3 in.
    Length (Metric) 76mm
    Length (Metric) Probe 7.6cm
    Material Stainless steel,Stainless steel
    Resolution 0.1°C
    Temperature Range (English) -58° to +392°F,-58° to +392°F
    Temperature Range (Metric) -50° to +200°C,-50° to +200°C
    For Use With Field studies, cuvettes, test tubes, flasks and beakers, reads temperature in sample reagents, water baths, semi solids and gases

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit