Learn More
H-B Instrument™ Durac™ Waterproof Electronic Thermometer
For reading temperatures in sample reagents, water baths, semi-solids and gases
Manufacturer:  H-B Instrument Company B609001800
 View more versions of this product
Catalog No. 13-201-770
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
2°C
Field studies, cuvettes, test tubes, flasks and beakers, reads temperature in sample reagents, water baths, semi solids and gases
4.9 in.
123mm
Stainless steel
-58° to +572°F
ISO/IEC 17025:2005, A2LA, NIST
Stainless Steel probe, probe cover with pocket clip, multilanguage instructions and statement of accuracy, LR44 battery
5 in.
12.3cm
0.1°C
-50° to +300°C