Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-B Instrument™ Easy-Read™ Environmentally Friendly Liquid-In-Glass Tank Thermometers

Cup holds liquid sample for effortless reading

Specifications

Immersion Type Partial
For Use With (Application) Ideal for use in chemical/petrochemical laboratories, food and beverage processing, dairies, breweries and distilleries.
Certifications/Compliance Traceable to NIST. Includes a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
Immersion Length 76 mm
View More Specs

Products 1
Catalog Number Mfr. No. Temperature Range (English) Temperature Range (Metric) Subdivision Material Price Quantity & Availability  
Catalog Number Mfr. No. Temperature Range (English) Temperature Range (Metric) Subdivision Material Price Quantity & Availability  
13-201-985
View Documents
H-B Instrument™
B603800300
n.a. -20°C to +110°C 1°C Glass N/A
Description

Description

  • 100% environmentally safe
  • Features a wood casing with brass cup to hold a liquid sample for a more accurate reading
  • Cup capacity: 20mL
  • Lead-free, yellow back glass and black liquid provides easy readability and fewer reading errors
  • Contains non-toxic, biodegradable, Enviro-Safe™ liquid
  • EnviroKleen™ certified

Applications:

Ideal for use in chemical/petrochemical laboratories, food and beverage processing, dairies, breweries and distilleries.

Order Info

Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.

Certifications

Manufactured and tested by our USA quality team, these thermometers are traceable to NIST™ and include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.

Specifications

Specifications

Partial
Ideal for use in chemical/petrochemical laboratories, food and beverage processing, dairies, breweries and distilleries.
Traceable to NIST. Includes a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
76 mm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit