Learn More
H-B™ Enviro-Safe™ General Purpose Liquid-In-Glass Thermometers, Partial Immersion
Certified environmentally safe thermometers replace dangerous mercury
Manufacturer:  H-B Instrument Company B605010200RMD
 View more versions of this product
Catalog No. 13-202-556
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
±2°F
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Partial
200mm
20° to 230°F
Individually serialized thermometers are traceable to NIST; include a multi-language Statement of Accuracy and Traceability.
50mm (2 in.)
7.9 in.
2°F
Not Applicable