Learn More
H-B Instrument™ Enviro-Safe™ General Purpose Liquid-In-Glass Thermometers, Partial Immersion 3 in.
Certified environmentally safe thermometers replace dangerous mercury
$36.65 - $43.45
Specifications
Certifications/Compliance Individually serialized thermometers are traceable to NIST; include a multi-language Statement of Accuracy and Traceability.
For Use With (Application) Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Immersion Type Partial
Immersion Length 76 mm
View More Specs
Products 9
Catalog Number Mfr. No. Temperature Range (English) Temperature Range (Metric) Subdivision Accuracy Price Quantity & Availability  
Catalog Number Mfr. No. Temperature Range (English) Temperature Range (Metric) Subdivision Accuracy Price Quantity & Availability  
13-202-561
View Documents
H-B Instrument™
B605020500RMD
0°F to 300°F 1°F ±2°F; ±3°F above 212°F
Each for $43.45
Add to cart
13-201-480
View Documents
H-B Instrument™
B605020000
-35°C to +50°C 1°C ±1°C
Each for $42.90
Add to cart
13-201-481
View Documents
H-B Instrument™
B605020300
-30°F to +120°F 1°F ±1°F
Each for $36.65
Add to cart
13-202-559
View Documents
H-B Instrument™
B605020200RMD
-20°C to +150°C 1°C ±1°C; ±1.5°C above 100°C This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-202-560
View Documents
H-B Instrument™
B605020400RMD
0°F to 230°F 2°F ±2°F This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-202-562
View Documents
H-B Instrument™
B605030000RMD
-10°C to +260°C 1°C ±1.5°C, ±2°C above 100°C, ±2.5°C above 200°C, ±3°C above 230°C This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-202-563
View Documents
H-B Instrument™
B605030100RMD
20°F to 500°F 2°F ±3°F, ±4°F above 212°F, ±5°F above 392°F This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-202-556
View Documents
H-B Instrument™
B605010200RMD
20°F to 230°F 2°F ±2°F This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-202-555
View Documents
H-B Instrument™
B605010100RMD
-10°C to +150°C 1°C ±1°C; ±1.5°C above 100°C This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Earth friendly, EnviroKleen™ certified thermometers
  • Feature green, non-toxic, biodegradable, Enviro-Safe™ liquid against lead-free, white back glass
  • Meet government, education, and industry mandates to purchase “Green” products
Applications:
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Order Info
Should you require an individualized Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.
Certifications
Individually serialized thermometers are traceable to NIST; include a multi-language Statement of Accuracy and Traceability.
Compliance
Manufactured and tested by USA quality team.
Specifications
Specifications
Individually serialized thermometers are traceable to NIST; include a multi-language Statement of Accuracy and Traceability.
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Partial
76 mm