Learn More
H-B™ Enviro-Safe™ General Purpose Liquid-In-Glass Thermometers, Total Immersion
Certified environmentally safe thermometers replace dangerous mercury
Manufacturer:  H-B Instrument Company B605070100RMD
 View more versions of this product
Catalog No. 13-202-573
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
±4°F
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
14 in.
2°F
Not Applicable
Individually serialized thermometers are traceable to NIST; include a multi-language Statement of Accuracy and Traceability.
Total
350mm
20° to 440°F