Learn More
H-B™ Enviro-Safe™ Environmentally Friendly Liquid-In-Glass Pocket Thermometers
Environmentally friendly portable thermometers
Manufacturer:  H-B Instrument Company B605701200RMD
 View more versions of this product
Catalog No. 13-202-578
  Check Availability
Add to cart
Specifications
Description
EnviroKleen™ certified thermometers feature green, nontoxic, biodegradable, Enviro-Safe liquid against lead-free, white back glass Total immersion pocket thermometers are a compact 160mm (6.3 in.) long Come with a case to protect from breakage Case options include: Plastic Window - encased in plastic with an open window Metal Window - encased in metal with an open window Closed Plastic - housed in removable plastic case Closed Metal - housed in a removable metal case Aluminum Duplex - encased in nickel-plated brass with an open window and housed in a removable aluminum case Nickel-Plated Brass Window - encased in nickel-plated brass with an open window
Description & Specifications
Specifications
±1°F
Ideal for field studies, waste water treatment and any “on-the-go” applications.
Window metal case
160mm
20° to 120°F
Manufactured and tested by USA quality team, and include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
Total
6.3 in.
1°F
Not Applicable