Learn More
H-B Instrument™ FRIO-Temp™ Mercury Oven Thermometers
Mercury fill, yellow back glass
Includes: Statement of Accuracy traceable to NIST (USA) and DKD (Europe), unique serial number, and hook-and-loop and magnet attachments.
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
13-202-634
View Documents
H-B Instrument™
20685RMD
N/A For Oven: 50deg. to 200deg.C This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Standard PTFE coating
  • Fine glass bead bottle fill
  • Accuracy: 1° or 2°C, depending on unit
  • Divisions: 1, 2 or 4, depending on unit