Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-B Instrument™ FRIO-Temp™ Precision Liquid-in-Glass Verification Thermometers Available on GSA/VA Contract for Federal Government customers only.

To verify sample temperatures in freezers, refrigerators, incubators and ovens with traceability to NIST™ and DAkkS.

Manufacturer:  H-B Instrument Company B610040200RMD

 View more versions of this product

Catalog No. 13-202-509


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

±1
Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries
Precision
98mm
0.5
10° to 45°C
Verification Thermometer
ISO/IEC 17025:2005, A2LA, NIST, DAkkS/PTB
Incubator
3.9 in.
Glass
Not Applicable
Enviro-Safe™
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit