Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-B Instrument™ Plastic Tube Storage Cases

With twist lock cap

$4.51 - $5.97

Specifications

Product Type Storage Case
For Use With (Application) Protect Glass thermometer while in storage
For Use With (Equipment) Thermometers
Material Plastic
View More Specs

Products 3
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity & Availability  
13-202-138
View Documents
H-B Instrument™
B614003600
12 in. 300 mm
Each for $5.97
Pack of 25 EA for $61.70
Add to cart
 
13-202-139
View Documents
H-B Instrument™
B614003700
16 in. 405 mm
Each for $4.51
Pack of 25 EA for $77.30
Add to cart
 
13-202-137
View Documents
H-B Instrument™
B614003500
6.3 in. 160 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Holds one thermometer
  • Can accomodate devices of length 6.3 to 16 in. (160 to 405mm)

Specifications

Specifications

Storage Case
Protect Glass thermometer while in storage
Thermometers
Plastic
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit