Learn More
H-B Instrument™ Enviro-Safe™ Environmentally Friendly Liquid-In-Glass Pocket Thermometers Available on GSA/VA Contract for Federal Government customers only.
Environmentally friendly portable thermometers
$25.25 - $35.60
Specifications
Certifications/Compliance Traceable to NIST. Includes a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
For Use With (Application) Ideal for field studies, waste water treatment and any “on-the-go” applications.
Immersion Type Total
Length (English) Overall 6.3 in.
Length (Metric) Overall 160 mm
View More Specs
Products 18
Catalog Number Mfr. No. Accuracy Includes Subdivision Price Quantity & Availability  
Catalog Number Mfr. No. Accuracy Includes Subdivision Price Quantity & Availability  
13-201-500
View Documents
H-B Instrument™
B605700400
±1°F Window plastic case 1°F
Each for $26.90
Add to cart
13-201-499
View Documents
H-B Instrument™
B605700300
±2°F Window plastic case 2°F
Each for $28.00
Add to cart
13-201-502
View Documents
H-B Instrument™
B605700600
±0.5°C Closed plastic case 0.5°C
Each for $25.25
Add to cart
13-201-504
View Documents
H-B Instrument™
B605700800
±1°F Closed plastic case 1°F
Each for $25.80
Add to cart
13-201-503
View Documents
H-B Instrument™
B605700700
±2°F Closed plastic case 2°F
Each for $28.25
Add to cart
13-201-501
View Documents
H-B Instrument™
B605700500
±1°C Closed plastic case 1°C
Each for $28.00
Add to cart
13-201-505
View Documents
H-B Instrument™
B605701800
±1°C Aluminum duplex case 1°C
Each for $35.30
Add to cart
13-201-506
View Documents
H-B Instrument™
B605701900
±0.5°C Aluminum duplex case 0.5°C
Each for $34.20
Add to cart
13-201-507
View Documents
H-B Instrument™
B605702000
±2°F Aluminum duplex case 2°F
Each for $35.30
Add to cart
13-201-508
View Documents
H-B Instrument™
B605702100
±1°F Aluminum duplex case 1°F
Each for $35.60
Add to cart
13-202-575
View Documents
H-B Instrument™
B605700900
±1°C Window metal case 1°C
Each for $34.50
Add to cart
13-202-574
View Documents
H-B Instrument™
B605700200
±0.5°C Window Plastic Case 0.5°C
Each for $28.50
Add to cart
13-202-578
View Documents
H-B Instrument™
B605701200RMD
±1°F Window metal case 1°F
Each for $27.15
Add to cart
13-202-576
View Documents
H-B Instrument™
B605701000
±0.5°C Window metal case 0.5°C
Each for $27.15
Add to cart
13-202-579
View Documents
H-B Instrument™
B605701300
±1°C Closed metal case 1°C
Each for $25.80
Add to cart
13-202-580 Available on GSA/VA Contract for Federal Government customers only.
View Documents
H-B Instrument™
B605701400
±0.5°C Closed metal case 0.5°C
Each for $27.45
Add to cart
13-202-581
View Documents
H-B Instrument™
B605701500
±2°F Closed metal case 2°F
Each for $28.25
Add to cart
13-201-498
View Documents
H-B Instrument™
B605700100
±1°C Window plastic case 1°C This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • EnviroKleen™ certified thermometers feature green, nontoxic, biodegradable, Enviro-Safe liquid against lead-free, white back glass
  • Total immersion pocket thermometers are a compact 160mm (6.3 in.) long
  • Come with a case to protect from breakage
Case options include:
Plastic Window - encased in plastic with an open window
Metal Window - encased in metal with an open window
Closed Plastic - housed in removable plastic case
Closed Metal - housed in a removable metal case
Aluminum Duplex - encased in nickel-plated brass with an open window and housed in a removable aluminum case
Nickel-Plated Brass Window - encased in nickel-plated brass with an open window
Applications:
Ideal for field studies, waste water treatment and any “on-the-go” applications.
Order Info
Should you require an individualized Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.
Certifications
Manufactured and tested by USA quality team and include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
Specifications
Specifications
Traceable to NIST. Includes a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
Total
160 mm
Ideal for field studies, waste water treatment and any “on-the-go” applications.
6.3 in.