Learn More
H-B Instrument™ Durac™ Plus™ Precision Liquid-In-Glass Partial Immersion Thermometers
Provides precision and ensures accuracy. Durac™ Plus™ precision thermometers are made with permanently fused markings and fractional degrees.
$110.00
Specifications
Certifications/Compliance ISO/IEC 17025:2005, A2LA, NIST
Material Glass
For Use With (Application) Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries
Grade Precision
Immersion Length 76 mm
View More Specs
Products 1
Catalog Number Mfr. No. Accuracy Subdivision Temperature Range (English) Length (English) Overall Price Quantity & Availability  
Catalog Number Mfr. No. Accuracy Subdivision Temperature Range (English) Length (English) Overall Price Quantity & Availability  
13-201-633
View Documents
H-B Instrument™
B607300300
±2°F 0.2°F 30°F to 124°F 17.9 in.
Each for $110.00
Add to cart
Description
Description
Durac™ Plus™ precision thermometers are made with permanently fused markings and fractional degrees to provide precision and ensure accuracy.
  • Mercury free, contain blue organic liquid against lead-free white back glass
  • Ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical/medical industry, manufacturing, dairies, breweries and distilleries
  • Traceable to NIST, individually serialized
  • Include multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
  • Tested and inspected by our USA quality team
Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.
Specifications
Specifications
ISO/IEC 17025:2005, A2LA, NIST
Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries
76 mm
Multilanguage instructions, statement of accuracy
Glass
Precision
Partial
Organic