Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-B Instrument™ Storage Tray for Angled Thermometer

Features a built-in angle to prevent separation of thermometer liquid

$90.20

Specifications

Material Polystyrene
For Use With (Equipment) Angled thermometer
Type Storage Tray
View More Specs

Products 2
Catalog Number Mfr. No. Depth (English) Length (English) Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Depth (English) Length (English) Width (English) Price Quantity & Availability  
13-202-140
View Documents
H-B Instrument™
B614005100RMD
1.2 in. 17.4 in. 13.4 in.
Each for $90.20
Add to cart
 
13-202-141
View Documents
H-B Instrument™
B614005300RMD
1.3 in. 17 in. 4 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Made from 2mm polystyrene
  • UV coated
  • Recyclable
  • Available in 3 or 14 slot versions with numbered slots

Specifications

Specifications

Polystyrene
Angled thermometer
Storage Tray
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit