Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-B Instrument™ Thermometer Cup Cases

With spring clip for holding device

Manufacturer:  H-B Instrument™ B614011100

 View more versions of this product

Catalog No. 13-202-145


Add to cart

Description

Description

  • Made of wood frame with brass cup or heat resistant black phenolic case
  • Cup capacity: 20mL
  • Thermometer length: 11.8 in. (300mm)
  • Case length: 15.9 in. (405mm)

Specifications

Specifications

11.8 in. Thermometer
405 mm
Cup Case
Brass cup
Brass
15.9 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit