Learn More
H-B Instrument™ Thermometer Magnifier
Readings appear three times larger than actual size
Manufacturer:  H-B Instrument™ B614007000RMD
Catalog No. 13-202-147
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Can snap on to thermometers of dimensions of 0.24 to 0.28 in. (6 to 7mm)
  • Dimension: 3.3 x 2.4 in. (82.6 x 60mm)
Specifications
Specifications
Thermometers
Readings appear 3X larger than actual size
Magnifier