Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-B Instrument™ Velvet Lined Thermometer Storage Cases

Single thermometer or nine thermometers can be stored according to choice

Specifications

Product Type Storage Case
For Use With (Application) Single thermometer can be stored
Material Velvet, Leatherette
View More Specs

Products 4
Catalog Number Mfr. No. For Use With (Equipment) Length (English) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Length (English) Length (Metric) Price Quantity & Availability  
13-201-518
View Documents
H-B Instrument™
B614003100
12 in. Thermometer 12 in. 305 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-201-519
View Documents
H-B Instrument™
B614003300
18 in. Thermometer 18 in. 460 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-202-135
View Documents
H-B Instrument™
B614003200
15.9 in. Thermometer 15.9 in. 405 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-202-136
View Documents
H-B Instrument™
B614003400
24 in. Thermometer 24 in. 610 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

 

  • Velvet lined
  • Leatherette
  • Holds one thermometer
  • Thermometer length range: 12 to 24 in. (305 to 610mm)

 

Specifications

Specifications

Storage Case
Single thermometer can be stored
Velvet, Leatherette
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit