Learn More
Chem-Impex International, Inc.Supplier Diversity Partner L-Cysteine, 52-90-4, MFCD00064306, 25g DFS Item
Manufacturer:  Chem-Impex International, Inc.Supplier Diversity Partner 0043725G
Molecular Formula C3H7NO2S, Purity Limit ≥ 99% (Titration: anhydrous basis), Molecular Weight 121.16, Melting Point 220°C (dec), Synonyms: L-Cys-OH, (R)-2-Amino-3-mercaptopropionic acid
Catalog No. 50-491-570