Learn More
Chem-Impex International, Inc.Supplier Diversity Partner Glycine tert-butyl ester hydrochloride, 27532-96-3, MFCD00058255, 5g DFS Item
Manufacturer:  Chem-Impex International, Inc.Supplier Diversity Partner 030725G
Molecular Formula C6H13NO2·HCl, Purity Limit ≥ 99% (HPLC), Molecular Weight 167.6, Melting Point 128-141 °C, Synonyms: Gly-OtBu·HCl, τ-Butyl glycinate hydrochloride
Catalog No. 50-497-031