Learn More
Chem-Impex International, Inc.Supplier Diversity Partner Nε-Boc-L-Lysine,2418-95-3,MFCD00037221,25G DFS Item
Manufacturer:  Chem-Impex International, Inc.Supplier Diversity Partner 0036325G
Molecular Formula C11H22N2O4; Purity Limit ≥ 98% (HPLC); Molecular Weight 246.30; Melting Point 192-202 °C; Storage Temp: Store at 0-8 °C; Synonyms: Boc-L-Lys-OH
Catalog No. 50-491-398